Assay Details
| Gene Symbol: RIMS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096989.1 | XP_020952648.1 | 1100 | 64 | ||
| XM_021096990.1 | XP_020952649.1 | 1210 | 64 | |||
| XM_021096991.1 | XP_020952650.1 | 1384 | 64 | |||
| XM_021096992.1 | XP_020952651.1 | 1245 | 64 | |||
| XM_021096993.1 | XP_020952652.1 | 1090 | 64 | |||
| XM_021096994.1 | XP_020952653.1 | 611 | 64 | |||
More Information
Set Membership: |
|

RIMS3
Genomic Map