Assay Details
| Gene Symbol: AIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122456.3 | XP_003122504.1 | 229 | 58 | ||
| XM_013994026.2 | XP_013849480.1 | 229 | 58 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: AIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122456.3 | XP_003122504.1 | 229 | 58 | ||
| XM_013994026.2 | XP_013849480.1 | 229 | 58 | |||
Set Membership: |
|