Assay Details
| Gene Symbol: UVSSA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021100878.1 | XP_020956537.1 | 622 | 66 | ||
| XM_021100879.1 | XP_020956538.1 | 686 | 66 | |||
| XM_021100880.1 | XP_020956539.1 | 1028 | 66 | |||
| XM_021100881.1 | XP_020956540.1 | 1133 | 66 | |||
| XM_021100882.1 | XP_020956541.1 | 590 | 66 | |||
| XM_021100883.1 | XP_020956542.1 | 624 | 66 | |||
More Information
Set Membership: |
|

UVSSA
Genomic Map