Assay Details
| Gene Symbol: LOC102158569 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013992987.2 | XP_013848441.1 | 1854 | 113 | ||
| XM_021095361.1 | XP_020951020.1 | 1854 | 113 | |||
| XM_021095379.1 | XP_020951038.1 | 2330 | 113 | |||
More Information
Set Membership: |
|

LOC102158569
Genomic Map