Assay Details
| Gene Symbol: TSKS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005664744.3 | XP_005664801.1 | 1382 | 90 | ||
| XM_021094748.1 | XP_020950407.1 | 1677 | 90 | |||
| XM_021094749.1 | XP_020950408.1 | 1649 | 90 | |||
| XM_021094750.1 | XP_020950409.1 | 1677 | 90 | |||
| XM_021094751.1 | XP_020950410.1 | 1649 | 90 | |||
| XM_021094752.1 | XP_020950411.1 | 1677 | 90 | |||
| XM_021094753.1 | XP_020950412.1 | 1676 | 90 | |||
More Information
Set Membership: |
|

TSKS
Genomic Map