Assay Details
| Gene Symbol: GYPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129173.4 | XP_003129221.1 | 497 | 90 | ||
| XM_005656526.3 | XP_005656583.1 | 419 | 90 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: GYPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129173.4 | XP_003129221.1 | 497 | 90 | ||
| XM_005656526.3 | XP_005656583.1 | 419 | 90 | |||
Set Membership: |
|