Assay Details
| Gene Symbol: ADNP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013993971.2 | XP_013849425.1 | 273 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013993971.2 transcripts.
Set Membership: |
|

ADNP2
Genomic Map