Assay Details
| Gene Symbol: PHF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095267.1 | XP_020950926.1 | 473 | 96 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PHF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095267.1 | XP_020950926.1 | 473 | 96 | ||