Assay Details
| Gene Symbol: PWWP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021073878.1 | XP_020929537.1 | 1295 | 109 | ||
| XM_021073880.1 | XP_020929539.1 | 1305 | 109 | |||
More Information
Set Membership: |
|

PWWP2B
Genomic Map