Assay Details
| Gene Symbol: FBXO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121138.4 | XP_003121186.1 | 1446 | 151 | ||
| XM_013992479.2 | XP_013847933.1 | 953 | 151 | |||
| XM_013992480.2 | XP_013847934.1 | 1440 | 151 | |||
| XM_013992481.2 | XP_013847935.1 | 1436 | 151 | |||
More Information
Set Membership: |
|

FBXO5
Genomic Map