Assay Details
| Gene Symbol: ZNF213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013995736.2 | XP_013851190.2 | 1437 | 67 | ||
| XM_013995737.2 | XP_013851191.2 | 1613 | 67 | |||
More Information
Set Membership: |
|

ZNF213
Genomic Map