Assay Details
| Gene Symbol: OCM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086087.1 | XP_020941746.1 | 671 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021086087.1 transcripts.
Set Membership: |
|

OCM
Genomic Map