Assay Details
| Gene Symbol: OCM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124292.3 | XP_003124340.1 | 825 | 104 | ||
| XM_021086086.1 | XP_020941745.1 | 825 | 104 | |||
| XM_021086087.1 | XP_020941746.1 | 739 | 104 | |||
More Information
Set Membership: |
|

OCM
Genomic Map