Assay Details
| Gene Symbol: RELCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121702.4 | XP_003121750.1 | 2152 | 65 | ||
| XM_005654481.3 | XP_005654538.1 | 2156 | 65 | |||
| XM_005654482.3 | XP_005654539.1 | 2078 | 65 | |||
| XM_021099712.1 | XP_020955371.1 | 2174 | 65 | |||
| XM_021099721.1 | XP_020955380.1 | 2174 | 65 | |||
| XM_021099741.1 | XP_020955400.1 | 2102 | 65 | |||
| XM_021099750.1 | XP_020955409.1 | 2174 | 65 | |||
More Information
Set Membership: |
|

RELCH
Genomic Map