Assay Details
| Gene Symbol: SEPTIN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005653856.3 | XP_005653913.2 | 1418 | 66 | ||
| GenBank mRNA | KU358542.1 | - | 6 - 7 | 1291 | 66 | |
More Information
Set Membership: |
|

SEPTIN9
Genomic Map