Assay Details
| Gene Symbol: LOC100620649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358454.3 | XP_003358502.1 | 320 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100620649| Gene Symbol: LOC100620649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358454.3 | XP_003358502.1 | 320 | 94 | ||