Assay Details
| Gene Symbol: TMEM255B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005653841.3 | XP_005653898.1 | 559 | 53 | ||
| XM_013978996.2 | XP_013834450.1 | 571 | 53 | |||
More Information
Set Membership: |
|

TMEM255B
Genomic Map