Assay Details
| Gene Symbol: AKAP17A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081363.1 | XP_020937022.1 | 931 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AKAP17A| Gene Symbol: AKAP17A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081363.1 | XP_020937022.1 | 931 | 89 | ||
Set Membership: |
|