Assay Details
| Gene Symbol: YPEL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132989.4 | XP_003133037.1 | 614 | 118 | ||
| XM_021072282.1 | XP_020927941.1 | 3601 | 118 | |||
| XM_021072284.1 | XP_020927943.1 | 246 | 118 | |||
More Information
Set Membership: |
|

YPEL1
Genomic Map