Assay Details
| Gene Symbol: DBF4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066859.1 | XP_020922518.1 | 856 | 77 | ||
| XM_021066860.1 | XP_020922519.1 | 856 | 77 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: DBF4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066859.1 | XP_020922518.1 | 856 | 77 | ||
| XM_021066860.1 | XP_020922519.1 | 856 | 77 | |||
Set Membership: |
|