Assay Details
| Gene Symbol: ASB15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013985647.2 | XP_013841101.1 | 1599 | 81 | ||
| XM_013985648.2 | XP_013841102.1 | 1537 | 81 | |||
| XM_013985649.2 | XP_013841103.1 | 1673 | 81 | |||
| XM_013985651.2 | XP_013841105.1 | 1813 | 81 | |||
| XM_013985654.2 | XP_013841108.1 | 1958 | 81 | |||
| XM_013985655.2 | XP_013841109.1 | 1534 | 81 | |||
| XM_013985656.2 | XP_013841110.1 | 1884 | 81 | |||
| XM_021079194.1 | XP_020934853.1 | 1518 | 81 | |||
| XM_021079195.1 | XP_020934854.1 | 2106 | 81 | |||
| XM_021079197.1 | XP_020934856.1 | 1530 | 81 | |||
More Information
Set Membership: |
|

ASB15
Genomic Map