Assay Details
| Gene Symbol: GPR174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013986259.2 | XP_013841713.1 | 544 | 95 | ||
| XM_013986260.2 | XP_013841714.1 | 544 | 95 | |||
| XM_013986261.2 | XP_013841715.1 | 386 | 95 | |||
| XM_013986262.2 | XP_013841716.1 | 349 | 95 | |||
| XM_021080858.1 | XP_020936517.1 | 349 | 95 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013986259.2, XM_013986260.2, XM_013986261.2, XM_013986262.2, XM_021080858.1 transcripts.
Set Membership: |
|

GPR174
Genomic Map