Assay Details
| Gene Symbol: GPR174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013986262.2 | XP_013841716.1 | 248 | 99 | ||
| XM_021080858.1 | XP_020936517.1 | 248 | 99 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013986262.2, XM_021080858.1 transcripts.
Set Membership: |
|

GPR174
Genomic Map