Assay Details
| Gene Symbol: ZNF276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003355714.4 | XP_003355762.1 | 1146 | 66 | ||
| XM_013994367.2 | XP_013849821.1 | 930 | 66 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ZNF276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003355714.4 | XP_003355762.1 | 1146 | 66 | ||
| XM_013994367.2 | XP_013849821.1 | 930 | 66 | |||
Set Membership: |
|