Assay Details
| Gene Symbol: SPATA31D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005660180.3 | XP_005660237.2 | 114 | 111 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005660180.3 transcripts.
Set Membership: |
|

SPATA31D1
Genomic Map