Assay Details
| Gene Symbol: PURG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003359494.4 | XP_003359542.1 | 151 | 76 | ||
| XM_005671806.3 | XP_005671863.1 | 171 | 76 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003359494.4, XM_005671806.3 transcripts.
Set Membership: |
|

PURG
Genomic Map