Assay Details
| Gene Symbol: RASSF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013989936.2 | XP_013845390.2 | 951 | 69 | ||
| XM_021082552.1 | XP_020938211.1 | 951 | 69 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RASSF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013989936.2 | XP_013845390.2 | 951 | 69 | ||
| XM_021082552.1 | XP_020938211.1 | 951 | 69 | |||
Set Membership: |
|