Assay Details
Gene Symbol: LOC100154415 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005670500.3 | XP_005670557.1 | 419 | 90 | ||
XM_013982727.2 | XP_013838181.1 | 567 | 90 | |||
XM_013982728.2 | XP_013838182.1 | 419 | 90 | |||
XM_013982729.2 | XP_013838183.1 | 267 | 90 | |||
XM_021072878.1 | XP_020928537.1 | 567 | 90 | |||
XM_021072880.1 | XP_020928539.1 | 258 | 90 | |||
XM_021072883.1 | XP_020928542.1 | 112 | 90 | |||
XM_021072884.1 | XP_020928543.1 | 567 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005670500.3, XM_013982728.2, XM_013982729.2, XM_021072880.1, XM_021072883.1, XM_021072884.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
