Assay Details
| Gene Symbol: DPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096832.1 | XP_020952491.1 | 129 | 85 | ||
| XM_021096833.1 | XP_020952492.1 | 2484 | 85 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: DPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096832.1 | XP_020952491.1 | 129 | 85 | ||
| XM_021096833.1 | XP_020952492.1 | 2484 | 85 | |||
Set Membership: |
|