Assay Details
| Gene Symbol: EIF2S3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134980.6 | XP_003135028.3 | 552 | 80 | ||
| XM_021080522.1 | XP_020936181.1 | 624 | 80 | |||
| GenBank mRNA | AK231727.1 | - | 1 - 2 | 92 | 80 | |
More Information
Set Membership: |
|

EIF2S3
Genomic Map