Assay Details
| Gene Symbol: MXI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133181.6 | XP_003133229.5 | 2714 | 77 | ||
| XM_021073198.1 | XP_020928857.1 | 2568 | 77 | |||
| XM_021073199.1 | XP_020928858.1 | 3269 | 77 | |||
| GenBank mRNA | AK239386.1 | - | 3 | 2024 | 77 | |
More Information
Set Membership: |
|

MXI1
Genomic Map