Assay Details
| Gene Symbol: ITPRIPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086544.1 | XP_020942203.1 | 1507 | 78 | ||
| GenBank mRNA | AK343596.1 | - | 1 | 923 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ITPRIPL2| Gene Symbol: ITPRIPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086544.1 | XP_020942203.1 | 1507 | 78 | ||
| GenBank mRNA | AK343596.1 | - | 1 | 923 | 78 | |
Set Membership: |
|