Assay Details
| Gene Symbol: JCAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003130706.6 | XP_003130754.3 | 3883 | 113 | ||
| XM_021064743.1 | XP_020920402.1 | 3895 | 113 | |||
| GenBank mRNA | AK394056.1 | - | 1 | 2024 | 113 | |
More Information
Set Membership: |
|

JCAD
Genomic Map