Assay Details
| Gene Symbol: VPS51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK231662.1 | - | 2 | 919 | 75 | |
| AK352378.1 | - | 2 | 897 | 75 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: VPS51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK231662.1 | - | 2 | 919 | 75 | |
| AK352378.1 | - | 2 | 897 | 75 | ||
Set Membership: |
|