Assay Details
| Gene Symbol: TMEM191C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005670922.3 | XP_005670979.1 | 1913 | 91 | ||
| XM_021073396.1 | XP_020929055.1 | 1913 | 91 | |||
More Information
Set Membership: |
|

TMEM191C
Genomic Map