Assay Details
| Gene Symbol: ENPP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927534.6 | XP_001927569.1 | 1759 | 64 | ||
| XM_005656228.3 | XP_005656285.1 | 1853 | 64 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ENPP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927534.6 | XP_001927569.1 | 1759 | 64 | ||
| XM_005656228.3 | XP_005656285.1 | 1853 | 64 | |||
Set Membership: |
|