Assay Details
| Gene Symbol: OGFOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244319.1 | NP_001231248.1 | 9 - 10 | 1076 | 61 | |
| XM_021093391.1 | XP_020949050.1 | 1029 | 61 | |||
| GenBank mRNA | AK232337.1 | - | 9 - 10 | 1076 | 61 | |
| AK389980.1 | - | 9 - 10 | 1077 | 61 | ||
More Information
Set Membership: |
|

OGFOD1
Genomic Map