Assay Details
| Gene Symbol: MSTO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244634.1 | NP_001231563.1 | 7 - 8 | 692 | 64 | |
| XM_005663292.3 | XP_005663349.1 | 64 | ||||
| XM_021088659.1 | XP_020944318.1 | 64 | ||||
| GenBank mRNA | AK347201.1 | - | 7 - 8 | 692 | 64 | |
More Information
Set Membership: |
|

MSTO1
Genomic Map