Assay Details
| Gene Symbol: PUS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244722.1 | NP_001231651.1 | 2 - 3 | 449 | 72 | |
| XM_005667485.3 | XP_005667542.1 | 590 | 72 | |||
| XM_021102025.1 | XP_020957684.1 | 685 | 72 | |||
| XM_021102026.1 | XP_020957685.1 | 526 | 72 | |||
| GenBank mRNA | AK230753.1 | - | 2 - 3 | 449 | 72 | |
More Information
Set Membership: |
|

PUS3
Genomic Map