Assay Details
| Gene Symbol: FOXO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214014.3 | NP_999179.3 | 2 - 3 | 2444 | 120 | |
| GenBank mRNA | AK234178.1 | - | 1 - 2 | 1012 | 120 | |
| AK234453.1 | - | 3 - 4 | 1428 | 120 | ||
| AY094061.1 | - | 2 - 3 | 2036 | 120 | ||
More Information
Set Membership: |
|

FOXO1
Genomic Map