Assay Details
| Gene Symbol: IFNAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204775.2 | NP_001191704.2 | 4 - 5 | 334 | 75 | |
| XM_021068450.1 | XP_020924109.1 | 573 | 75 | |||
| XM_021068451.1 | XP_020924110.1 | 574 | 75 | |||
| XM_021068452.1 | XP_020924111.1 | 490 | 75 | |||
| XM_021068453.1 | XP_020924112.1 | 463 | 75 | |||
| GenBank mRNA | AK231609.1 | - | 5 - 6 | 432 | 75 | |
| HQ603084.1 | - | 4 - 5 | 334 | 75 | ||
| HQ665551.1 | - | 3 - 4 | 219 | 75 | ||
More Information
Set Membership: |
|

IFNAR2
Genomic Map