Assay Details
| Gene Symbol: DDIT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243452.1 | NP_001230381.1 | 2 - 3 | 411 | 55 | |
| GenBank mRNA | AK392742.1 | - | 2 - 3 | 410 | 55 | |
| CJ020439.1 | - | 2 - 3 | 411 | 55 | ||
More Information
Set Membership: |
|

DDIT4
Genomic Map