Assay Details
Gene Symbol: RBMX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243671.1 | NP_001230600.1 | 1 - 2 | 37 | 71 | |
GenBank mRNA | AK240590.1 | - | 1 - 2 | 37 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001243671.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
