Assay Details
Gene Symbol: RBMX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243671.1 | NP_001230600.1 | 3 - 4 | 206 | 100 | |
GenBank mRNA | AK240590.1 | - | 3 - 4 | 206 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
