Assay Details
| Gene Symbol: ACT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF369524.1 | - | 1 - 2 | 98 | ||
| AF369525.1 | - | 1 - 2 | 182 | 98 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACT1| Gene Symbol: ACT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF369524.1 | - | 1 - 2 | 98 | ||
| AF369525.1 | - | 1 - 2 | 182 | 98 | ||
Set Membership: |
|