Assay Details
| Gene Symbol: CIPK04 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | FJ901232.1 | - | 6 - 7 | 531 | 84 | |
| FR669159.1 | - | 5 - 6 | 504 | 84 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CIPK04| Gene Symbol: CIPK04 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | FJ901232.1 | - | 6 - 7 | 531 | 84 | |
| FR669159.1 | - | 5 - 6 | 504 | 84 | ||
Set Membership: |
|