Assay Details
| Gene Symbol: CIPK04 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | FJ901232.1 | - | 8 - 9 | 711 | 71 | |
| FR669159.1 | - | 7 - 8 | 684 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CIPK04| Gene Symbol: CIPK04 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | FJ901232.1 | - | 8 - 9 | 711 | 71 | |
| FR669159.1 | - | 7 - 8 | 684 | 71 | ||
Set Membership: |
|