Assay Details
| Gene Symbol: tram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001210.2 | NP_001001210.1 | 10 - 11 | 1131 | 68 | |
| GenBank mRNA | BC067965.2 | - | 10 - 11 | 1131 | 68 | |
| CR760366.2 | - | 10 - 11 | 1099 | 68 | ||
More Information
Set Membership: |
|

tram1
Genomic Map