Assay Details
| Gene Symbol: prss29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001228.1 | NP_001001228.1 | 4 - 5 | 681 | 77 | |
| GenBank mRNA | BC067937.1 | - | 4 - 5 | 681 | 77 | |
More Information
Set Membership: |
|

prss29
Genomic Map