Assay Details
| Gene Symbol: itm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004833.1 | NP_001004833.1 | 1 - 2 | 170 | 79 | |
| GenBank mRNA | BC074629.1 | - | 1 - 2 | 170 | 79 | |
More Information
Set Membership: |
|

itm2a
Genomic Map